Product Details
- SNP ID
-
rs7723575
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:178775016 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTCGCAGACTGAGCAGACAATGCTT[C/T]CTCAAAGGGGATCTTCAGGCTCGGG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AACSP1
PubMed Links
Gene Details
- Gene
- AACSP1
- Gene Name
- acetoacetyl-CoA synthetase pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details