Product Details

SNP ID
rs17856609
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:112406882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGAGGCACATCTTCAGTAGGATA[A/C]TATCTAGGCTGTGGAGAGTCCATAG
Phenotype
MIM: 603703
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RPL6 PubMed Links

Gene Details

Gene
RPL6
Gene Name
ribosomal protein L6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000970.4 624 Intron NP_000961.2
NM_001024662.2 624 Intron NP_001019833.1
NM_001320137.1 624 Intron NP_001307066.1
NM_001320138.1 624 Nonsense Mutation TAG,TAT *115Y NP_001307067.1
NM_001320139.1 624 Intron NP_001307068.1
NM_001320140.1 624 Nonsense Mutation TAG,TAT *115Y NP_001307069.1
NM_001320141.1 624 Nonsense Mutation TAG,TAT *115Y NP_001307070.1
NM_001320142.1 624 Intron NP_001307071.1
XM_017019781.1 624 Nonsense Mutation TAG,TAT *115Y XP_016875270.1
XM_017019782.1 624 Intron XP_016875271.1

View Full Product Details