Product Details

SNP ID
rs17853695
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:57224986 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAGCTCCCAGAGCCAAGGTCT[A/T]CGGACGGCCTAGGCGTGGGCCGCGC
Phenotype
MIM: 604637 MIM: 138450
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NDUFA4L2 PubMed Links

Gene Details

Gene
NDUFA4L2
Gene Name
NDUFA4, mitochondrial complex associated like 2
There are no transcripts associated with this gene.

Gene
NXPH4
Gene Name
neurexophilin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007224.3 341 Missense Mutation ACG,TCG T56S NP_009155.1
XM_017018747.1 341 Missense Mutation ACG,TCG T56S XP_016874236.1
Gene
SHMT2
Gene Name
serine hydroxymethyltransferase 2
There are no transcripts associated with this gene.

View Full Product Details