Product Details
- SNP ID
-
rs7577689
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:69459154 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTTGACCCTGCATTCCGGGAAA[A/G]GTACCTCTGGGTTGTCACTACAGAG
- Phenotype
-
MIM: 616405
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AAK1
PubMed Links
Gene Details
- Gene
- AAK1
- Gene Name
- AP2 associated kinase 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_014911.3 |
20106 |
UTR 3 |
|
|
NP_055726.3 |
View Full Product Details