Product Details

SNP ID
rs17294478
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:183806265 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCATCTGTCAGTGTAAAATGACTTG[C/T]TTCTGTAAGCAGGATGTCACCTCCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
STOX2 PubMed Links

Gene Details

Gene
STOX2
Gene Name
storkhead box 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020225.2 Intron NP_064610.1
XM_011532129.1 Intron XP_011530431.1
XM_011532130.1 Intron XP_011530432.1
XM_011532131.1 Intron XP_011530433.1
XM_017008466.1 Intron XP_016863955.1
XM_017008467.1 Intron XP_016863956.1

View Full Product Details