Product Details

SNP ID
rs17850934
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55819407 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGAGACTCCTGACCAAGGAAAGGC[C/T]CGGCTACTGACACACTGGGAGCAAA
Phenotype
MIM: 606092 MIM: 610074 MIM: 610049
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC14 PubMed Links

Gene Details

Gene
DNAJC14
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
There are no transcripts associated with this gene.

Gene
ORMDL2
Gene Name
ORMDL sphingolipid biosynthesis regulator 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014182.4 332 Silent Mutation GCC,GCT A80A NP_054901.1
Gene
SARNP
Gene Name
SAP domain containing ribonucleoprotein
There are no transcripts associated with this gene.

View Full Product Details