Product Details

SNP ID
rs17119598
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96820578 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGAAGGAGGAGTTCCCGAGATG[C/T]GGAATGTCCTGGAAGCTAAGGGAAG
Phenotype
MIM: 607804 MIM: 607805
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CNNM3 PubMed Links
Additional Information
For this assay, SNP(s) [rs28438599] are located under a probe and SNP(s) [rs75853489] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CNNM3
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017623.4 Intron NP_060093.3
NM_199078.2 Intron NP_951060.1
XM_011510957.2 Intron XP_011509259.1
XM_017003800.1 Intron XP_016859289.1
Gene
CNNM4
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 4
There are no transcripts associated with this gene.

View Full Product Details