Product Details

SNP ID
rs7501705
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18381197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTTTGCTGGGCTCCCCTGTGT[C/G]TTTGTCACCTGCCTCATGGACGCCC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
EVPLL PubMed Links

Gene Details

Gene
EVPLL
Gene Name
envoplakin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145127.1 Intron NP_001138599.1
XM_011523982.2 Intron XP_011522284.1
XM_011523983.2 Intron XP_011522285.1
XM_011523985.2 Intron XP_011522287.1
XM_011523986.2 Intron XP_011522288.1
XM_011523987.2 Intron XP_011522289.1

View Full Product Details