Product Details

SNP ID
rs16887728
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38733954 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGAATTCGGGTCCTCCTTAGAC[A/G]TCCCTGCCTCAAATAATGCACTGTA
Phenotype
MIM: 605301
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TACC1 PubMed Links

Gene Details

Gene
TACC1
Gene Name
transforming acidic coiled-coil containing protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122824.1 Intron NP_001116296.1
NM_001146216.2 Intron NP_001139688.1
NM_006283.2 Intron NP_006274.2
XM_005273625.4 Intron XP_005273682.1
XM_005273628.1 Intron XP_005273685.1
XM_005273629.1 Intron XP_005273686.1
XM_005273630.1 Intron XP_005273687.1
XM_011544631.1 Intron XP_011542933.1
XM_011544632.2 Intron XP_011542934.1
XM_011544633.1 Intron XP_011542935.1
XM_011544634.1 Intron XP_011542936.1
XM_011544635.1 Intron XP_011542937.1
XM_011544636.2 Intron XP_011542938.1
XM_011544637.1 Intron XP_011542939.1
XM_011544638.2 Intron XP_011542940.1
XM_017013766.1 Intron XP_016869255.1
XM_017013767.1 Intron XP_016869256.1
XM_017013768.1 Intron XP_016869257.1
XM_017013769.1 Intron XP_016869258.1
XM_017013770.1 Intron XP_016869259.1
XM_017013771.1 Intron XP_016869260.1
XM_017013772.1 Intron XP_016869261.1
XM_017013773.1 Intron XP_016869262.1
XM_017013774.1 Intron XP_016869263.1
XM_017013775.1 Intron XP_016869264.1
XM_017013776.1 Intron XP_016869265.1
XM_017013777.1 Intron XP_016869266.1
XM_017013778.1 Intron XP_016869267.1

View Full Product Details