Product Details

SNP ID
rs17303905
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:65658473 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAGTCTATTTCCAATATTTATCT[A/G]GGTTTCTCTTCTACCAGCTGCCAAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MTFR1 PubMed Links

Gene Details

Gene
MTFR1
Gene Name
mitochondrial fission regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145838.1 Intron NP_001139310.1
NM_001145839.1 Intron NP_001139311.1
NM_014637.3 Intron NP_055452.3
XM_006716484.2 Intron XP_006716547.2
XM_011517626.1 Intron XP_011515928.1
XM_011517627.2 Intron XP_011515929.1
XM_011517628.2 Intron XP_011515930.1

View Full Product Details