Product Details

SNP ID
rs34268073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:49489252 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGGCAGGCCCCGGGACATACTGGT[C/T]CCATCGACAGTCCCAGCACTGCCCA
Phenotype
MIM: 604557
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF423 PubMed Links

Gene Details

Gene
ZNF423
Gene Name
zinc finger protein 423
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271620.2 6373 UTR 3 NP_001258549.1
NM_015069.4 6373 UTR 3 NP_055884.2
XM_005255856.4 6373 Intron XP_005255913.1
XM_005255857.4 6373 Intron XP_005255914.1
XM_006721171.3 6373 UTR 3 XP_006721234.1
XM_017023076.1 6373 UTR 3 XP_016878565.1
XM_017023077.1 6373 Intron XP_016878566.1
XM_017023078.1 6373 Intron XP_016878567.1

View Full Product Details