Product Details

SNP ID
rs35504571
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:42746006 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGGACGAACTCCTCTGGTCATGG[C/T]GGGGCCGTCCAGCCCTAGATGGTGA
Phenotype
MIM: 607465 MIM: 611695
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDAN1 PubMed Links

Gene Details

Gene
CDAN1
Gene Name
codanin 1
There are no transcripts associated with this gene.

Gene
TTBK2
Gene Name
tau tubulin kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173500.3 3845 Missense Mutation CAC,CGC H1175R NP_775771.3
XM_005254171.4 3845 Missense Mutation CAC,CGC H1181R XP_005254228.1
XM_005254173.4 3845 Missense Mutation CAC,CGC H1106R XP_005254230.1
XM_006720402.3 3845 Missense Mutation CAC,CGC H1170R XP_006720465.1
XM_006720403.3 3845 Missense Mutation CAC,CGC H1106R XP_006720466.1
XM_017021950.1 3845 Missense Mutation CAC,CGC H1082R XP_016877439.1

View Full Product Details