Product Details

SNP ID
rs17243515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:43192319 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCGGCGCCCTCACCTCGGGACATC[A/C]ACACACCGACCGCTCCTGCTCCAGA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NIM1K PubMed Links

Gene Details

Gene
NIM1K
Gene Name
NIM1 serine/threonine protein kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153361.3 252 UTR 5 NP_699192.1
XM_006714450.1 252 Intron XP_006714513.1
XM_006714451.1 252 UTR 5 XP_006714514.1
XM_017009164.1 252 UTR 5 XP_016864653.1
XM_017009165.1 252 Intron XP_016864654.1

View Full Product Details