Product Details

SNP ID
rs16938887
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74352800 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTGAGCTTTTTGGAAATGGATAAG[A/T]GTGTTAATTTGTATTTCTTGGGGGA
Phenotype
MIM: 606598
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GDAP1 PubMed Links

Gene Details

Gene
GDAP1
Gene Name
ganglioside induced differentiation associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040875.2 Intron NP_001035808.1
NM_018972.2 Intron NP_061845.2
XM_011517551.2 Intron XP_011515853.1
XM_011517552.2 Intron XP_011515854.1
XM_017013585.1 Intron XP_016869074.1
XM_017013586.1 Intron XP_016869075.1

View Full Product Details