Product Details

SNP ID
rs17352788
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123853642 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAGGGGTAGGTACGCCTCACCAC[A/G]GGCAAGCGTGGAGTAACATATTTTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FER1L6 PubMed Links

Gene Details

Gene
FER1L6
Gene Name
fer-1 like family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039112.2 Intron NP_001034201.2
XM_006716618.3 Intron XP_006716681.1
XM_011517231.2 Intron XP_011515533.1
XM_011517232.2 Intron XP_011515534.1
XM_011517233.2 Intron XP_011515535.1
XM_011517235.2 Intron XP_011515537.1
XM_011517236.2 Intron XP_011515538.1

View Full Product Details