Product Details
- SNP ID
-
rs17098687
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:87819842 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GACCATGAAATGCCAAAATCACAAG[C/G]CTTTAAAGGATGGGATGAAAGGGAT
- Phenotype
-
MIM: 614452
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ATAD1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138125275] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ATAD1
- Gene Name
- ATPase family, AAA domain containing 1
- Gene
- CFL1P1
- Gene Name
- cofilin 1 pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details