Product Details

SNP ID
rs17432442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:88717740 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTCCTGCTAATATCTCATCTGGA[A/T]ATATATAGAAATATATTACTCAAGA
Phenotype
MIM: 613922
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LIPK PubMed Links

Gene Details

Gene
LIPK
Gene Name
lipase family member K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080518.1 Intron NP_001073987.1
XM_011540073.2 Intron XP_011538375.1
XM_011540075.1 Intron XP_011538377.1
XM_011540077.1 Intron XP_011538379.1
XM_011540082.1 Intron XP_011538384.1
XM_017016545.1 Intron XP_016872034.1
XM_017016546.1 Intron XP_016872035.1
XM_017016547.1 Intron XP_016872036.1
XM_017016548.1 Intron XP_016872037.1

View Full Product Details