Product Details

SNP ID
rs17057269
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:27503812 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCAGGCCGAACCACCCAGAACCCT[C/T]GGGAGCATTAGGTCAGAATCCATTG
Phenotype
MIM: 132811
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHX2 PubMed Links

Gene Details

Gene
EPHX2
Gene Name
epoxide hydrolase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256482.1 Intron NP_001243411.1
NM_001256483.1 Intron NP_001243412.1
NM_001256484.1 Intron NP_001243413.1
NM_001979.5 Intron NP_001970.2
XM_017013199.1 Intron XP_016868688.1
XM_017013200.1 Intron XP_016868689.1

View Full Product Details