Product Details

SNP ID
rs17704117
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:140128691 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTGAAGTATTGTGATTATTTTAGG[C/T]ATTGATTCTTACAAAATATATACTG
Phenotype
MIM: 600473
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IGIP PubMed Links

Gene Details

Gene
IGIP
Gene Name
IgA inducing protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007189.1 2756 UTR 3 NP_001007190.1
Gene
PURA
Gene Name
purine rich element binding protein A
There are no transcripts associated with this gene.

View Full Product Details