Product Details

SNP ID
rs17367317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:84598162 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAAAGAAAAAGAAAAAGAAAACCA[C/T]GAAGAAACAGACAGAAAATGTTGAA
Phenotype
MIM: 603548
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDS1 PubMed Links

Gene Details

Gene
CDS1
Gene Name
CDP-diacylglycerol synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001263.3 Intron NP_001254.2
XM_005262687.2 Intron XP_005262744.1
XM_017007648.1 Intron XP_016863137.1
XM_017007649.1 Intron XP_016863138.1
XM_017007650.1 Intron XP_016863139.1
XM_017007651.1 Intron XP_016863140.1

View Full Product Details