Product Details

SNP ID
rs17128155
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:85731581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTAAGTCTTTTAAGCAATATTTAT[C/T]GAATACTAAGTATATTTAGCATAGT
Phenotype
MIM: 610025
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL24A1 PubMed Links

Gene Details

Gene
COL24A1
Gene Name
collagen type XXIV alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152890.5 Intron NP_690850.2
XM_017000924.1 Intron XP_016856413.1
XM_017000925.1 Intron XP_016856414.1
XM_017000926.1 Intron XP_016856415.1
XM_017000927.1 Intron XP_016856416.1
XM_017000928.1 Intron XP_016856417.1
XM_017000929.1 Intron XP_016856418.1
XM_017000930.1 Intron XP_016856419.1

View Full Product Details