Product Details

SNP ID
rs16915859
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:86575026 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGATAAGTGTCACTTTAGCCTGT[A/G]TCCTCCAGTCCAAGTCCCTTGAGCC
Phenotype
MIM: 605080
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CNGB3 PubMed Links

Gene Details

Gene
CNGB3
Gene Name
cyclic nucleotide gated channel beta 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019098.4 3256 UTR 3 NP_061971.3
XM_011517138.2 3256 UTR 3 XP_011515440.1

View Full Product Details