Product Details
- SNP ID
-
rs17021369
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:28325478 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGATACAAAGGTCAGGAGAAAGGT[G/T]GTTCTAGTCTCATCTCTGTACTCCT
- Phenotype
-
MIM: 609916
MIM: 615166
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
AZI2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76430272] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AZI2
- Gene Name
- 5-azacytidine induced 2
- Gene
- CMC1
- Gene Name
- C-X9-C motif containing 1
There are no transcripts associated with this gene.
View Full Product Details