Product Details

SNP ID
rs17852683
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:51895282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCAGGGTGCCCCGCCACCAGGCCT[G/T]GATCTTGGTGGCTACCGTATCTTTG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
IQCF1 PubMed Links

Gene Details

Gene
IQCF1
Gene Name
IQ motif containing F1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152397.2 324 Missense Mutation AAG,CAG K76Q NP_689610.2
XM_011533366.2 324 Missense Mutation AAG,CAG K41Q XP_011531668.1
XM_017005729.1 324 Missense Mutation AAG,CAG K41Q XP_016861218.1

View Full Product Details