Product Details

SNP ID
rs7061079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106696338 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTTTAAGACCTGCCAAATAGTGTG[C/T]CATTGGATTTTTTCCTTCCTAGAAA
Phenotype
MIM: 300439
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RNF128 PubMed Links

Gene Details

Gene
RNF128
Gene Name
ring finger protein 128, E3 ubiquitin protein ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024539.3 Intron NP_078815.3
NM_194463.1 Intron NP_919445.1

View Full Product Details