Product Details

SNP ID
rs7880561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106913004 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAAAGCGAGTCAGCAATGGGAA[G/T]ATCTGCATAAGTGGGTGGACCGATA
Phenotype
MIM: 300520 MIM: 300575
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLDN2 PubMed Links

Gene Details

Gene
CLDN2
Gene Name
claudin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171092.1 Intron NP_001164563.1
NM_001171095.1 Intron NP_001164566.1
NM_020384.3 Intron NP_065117.1
Gene
RIPPLY1
Gene Name
ripply transcriptional repressor 1
There are no transcripts associated with this gene.

View Full Product Details