Product Details

SNP ID
rs121907966
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:72345477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTCACAGCGGAAGTGTGACAAAC[G/A]TTCATAGGCAAATGTCAGGTCAGAT
Phenotype
MIM: 606869
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
HEXA PubMed Links

Gene Details

Gene
HEXA
Gene Name
hexosaminidase subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000520.5 2029 Missense Mutation CGT,TGT R499C NP_000511.2
NM_001318825.1 2029 Missense Mutation CGT,TGT R510C NP_001305754.1

View Full Product Details