Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_012164.3 | 7863 | UTR 3 | NP_036296.2 | ||
XM_005251910.3 | 7863 | Intron | XP_005251967.1 | ||
XM_005251913.2 | 7863 | Intron | XP_005251970.1 | ||
XM_006717049.3 | 7863 | Intron | XP_006717112.1 | ||
XM_006717051.2 | 7863 | Intron | XP_006717114.1 | ||
XM_011518518.1 | 7863 | Intron | XP_011516820.1 | ||
XM_017014615.1 | 7863 | Intron | XP_016870104.1 | ||
XM_017014616.1 | 7863 | Intron | XP_016870105.1 | ||
XM_017014617.1 | 7863 | Intron | XP_016870106.1 |