Product Details
- SNP ID
-
rs17855660
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:136672078 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTTTCTTCAATGATGAGTTTCTCTT[A/C]CTCAGGGAGTAGGACTTGTGGAATG
- Phenotype
-
MIM: 300267
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF6
PubMed Links
Gene Details
- Gene
- ARHGEF6
- Gene Name
- Rac/Cdc42 guanine nucleotide exchange factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306177.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E539* |
NP_001293106.1 |
NM_004840.2 |
2944 |
Nonsense Mutation |
GAA,TAA |
E693* |
NP_004831.1 |
XM_005262499.2 |
2944 |
Nonsense Mutation |
GAA,TAA |
E644* |
XP_005262556.1 |
XM_011531412.2 |
2944 |
Nonsense Mutation |
GAA,TAA |
E720* |
XP_011529714.1 |
XM_011531413.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E697* |
XP_011529715.1 |
XM_011531414.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E690* |
XP_011529716.1 |
XM_011531415.2 |
2944 |
Nonsense Mutation |
GAA,TAA |
E663* |
XP_011529717.1 |
XM_011531416.2 |
2944 |
Nonsense Mutation |
GAA,TAA |
E566* |
XP_011529718.1 |
XM_011531417.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E543* |
XP_011529719.1 |
XM_017029975.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E724* |
XP_016885464.1 |
XM_017029976.1 |
2944 |
Nonsense Mutation |
GAA,TAA |
E490* |
XP_016885465.1 |
View Full Product Details