Product Details

SNP ID
rs17831942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:20665426 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TAATGAATGACGAATCAGCTAATTC[A/G]TCTCTTTATCTCATTCTATAATAAA
Phenotype
MIM: 614606
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOCAD PubMed Links

Gene Details

Gene
FOCAD
Gene Name
focadhesin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017794.4 Intron NP_060264.4
XM_005251494.3 Intron XP_005251551.2
XM_011517945.2 Intron XP_011516247.1
XM_017014852.1 Intron XP_016870341.1
XM_017014853.1 Intron XP_016870342.1
XM_017014854.1 Intron XP_016870343.1
XM_017014855.1 Intron XP_016870344.1
XM_017014856.1 Intron XP_016870345.1
XM_017014857.1 Intron XP_016870346.1
XM_017014858.1 Intron XP_016870347.1
XM_017014859.1 Intron XP_016870348.1

View Full Product Details