Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_000053.3 | 5642 | UTR 3 | NP_000044.2 | ||
NM_001005918.2 | 5642 | UTR 3 | NP_001005918.1 | ||
NM_001243182.1 | 5642 | UTR 3 | NP_001230111.1 | ||
XM_005266423.2 | 5642 | UTR 3 | XP_005266480.1 | ||
XM_005266424.4 | 5642 | UTR 3 | XP_005266481.1 | ||
XM_005266427.2 | 5642 | UTR 3 | XP_005266484.1 | ||
XM_005266428.1 | 5642 | UTR 3 | XP_005266485.1 | ||
XM_005266430.4 | 5642 | UTR 3 | XP_005266487.1 | ||
XM_005266431.3 | 5642 | UTR 3 | XP_005266488.1 | ||
XM_005266432.2 | 5642 | UTR 3 | XP_005266489.1 | ||
XM_006719837.3 | 5642 | UTR 3 | XP_006719900.1 | ||
XM_006719838.1 | 5642 | UTR 3 | XP_006719901.1 | ||
XM_006719839.1 | 5642 | UTR 3 | XP_006719902.1 | ||
XM_011535117.2 | 5642 | UTR 3 | XP_011533419.1 | ||
XM_011535118.1 | 5642 | UTR 3 | XP_011533420.1 | ||
XM_011535119.1 | 5642 | UTR 3 | XP_011533421.1 | ||
XM_011535122.2 | 5642 | UTR 3 | XP_011533424.1 | ||
XM_017020627.1 | 5642 | UTR 3 | XP_016876116.1 | ||
XM_017020628.1 | 5642 | UTR 3 | XP_016876117.1 |