Product Details

SNP ID
rs16893887
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:28267053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCTCAGCAGGCAGCACCCATGTT[A/T]GTCTCCTTCTGCGATGCCTAAGCGG
Phenotype
MIM: 611643 MIM: 616474
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
NKAPL PubMed Links

Gene Details

Gene
NKAPL
Gene Name
NFKB activating protein like
There are no transcripts associated with this gene.

Gene
ZKSCAN4
Gene Name
zinc finger with KRAB and SCAN domains 4
There are no transcripts associated with this gene.

Gene
ZSCAN26
Gene Name
zinc finger and SCAN domain containing 26
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001023560.3 44 UTR 5 NP_001018854.2
NM_001111039.2 44 UTR 5 NP_001104509.1
NM_001287421.1 44 Intron NP_001274350.1
NM_001287422.1 44 UTR 5 NP_001274351.1
NM_152736.5 44 UTR 5 NP_689949.3
XM_011514862.2 44 UTR 5 XP_011513164.1
XM_011514864.1 44 Intron XP_011513166.1
XM_011514866.1 44 Intron XP_011513168.1
XM_011514867.1 44 Intron XP_011513169.1
XM_011514869.1 44 Intron XP_011513171.1
XM_017011264.1 44 UTR 5 XP_016866753.1
XM_017011265.1 44 Intron XP_016866754.1

View Full Product Details