Product Details
- SNP ID
-
rs17041314
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:100170847 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATACAAGGATTTCTACATTCTGAAT[A/G]TGAACCTTTGGTAGAAAAGCCAAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GOLGA2P5
PubMed Links
Gene Details
- Gene
- GOLGA2P5
- Gene Name
- golgin A2 pseudogene 5
There are no transcripts associated with this gene.
View Full Product Details