Product Details
- SNP ID
-
rs17861296
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:90348731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAATTTCTGTCATGCTTTTGCTTTT[C/T]TCTTTCCCCCAAAAAAATGTTTCTA
- Phenotype
-
MIM: 610920
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GTPBP10
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56837109] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GTPBP10
- Gene Name
- GTP binding protein 10
- Gene
- LOC101927446
- Gene Name
- uncharacterized LOC101927446
There are no transcripts associated with this gene.
View Full Product Details