Product Details
- SNP ID
-
rs17865323
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:90245460 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGAAGCCGGGGCGACCGCCGAGGC[C/T]CAGGAATCCGGCATCAGGAACAAGT
- Phenotype
-
MIM: 605094
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CFAP69
PubMed Links
Gene Details
- Gene
- CFAP69
- Gene Name
- cilia and flagella associated protein 69
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001039706.2 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
NP_001034795.2 |
NM_001160138.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
NP_001153610.1 |
XM_005250600.3 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_005250657.1 |
XM_011516577.2 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_011514879.1 |
XM_011516578.2 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_011514880.1 |
XM_011516579.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_011514881.1 |
XM_017012624.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868113.1 |
XM_017012625.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868114.1 |
XM_017012626.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868115.1 |
XM_017012627.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868116.1 |
XM_017012628.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868117.1 |
XM_017012629.1 |
266 |
UTR 5 |
|
|
XP_016868118.1 |
XM_017012630.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868119.1 |
XM_017012631.1 |
266 |
Intron |
|
|
XP_016868120.1 |
XM_017012632.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868121.1 |
XM_017012633.1 |
266 |
Intron |
|
|
XP_016868122.1 |
XM_017012634.1 |
266 |
Intron |
|
|
XP_016868123.1 |
XM_017012635.1 |
266 |
Intron |
|
|
XP_016868124.1 |
XM_017012636.1 |
266 |
Intron |
|
|
XP_016868125.1 |
XM_017012637.1 |
266 |
Intron |
|
|
XP_016868126.1 |
XM_017012638.1 |
266 |
Intron |
|
|
XP_016868127.1 |
XM_017012639.1 |
266 |
Intron |
|
|
XP_016868128.1 |
XM_017012640.1 |
266 |
Intron |
|
|
XP_016868129.1 |
XM_017012641.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868130.1 |
XM_017012642.1 |
266 |
Silent Mutation |
GCC,GCT |
A12A |
XP_016868131.1 |
- Gene
- STEAP2
- Gene Name
- STEAP2 metalloreductase
There are no transcripts associated with this gene.
View Full Product Details