Product Details

SNP ID
rs17867394
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:90247525 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTATCAAATGGAGATAATGACAGT[A/C]CTCATGTCCATTGTAAAGATTAAAT
Phenotype
MIM: 605094
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
CFAP69 PubMed Links

Gene Details

Gene
CFAP69
Gene Name
cilia and flagella associated protein 69
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039706.2 Intron NP_001034795.2
NM_001160138.1 Intron NP_001153610.1
XM_005250600.3 Intron XP_005250657.1
XM_011516577.2 Intron XP_011514879.1
XM_011516578.2 Intron XP_011514880.1
XM_011516579.1 Intron XP_011514881.1
XM_017012624.1 Intron XP_016868113.1
XM_017012625.1 Intron XP_016868114.1
XM_017012626.1 Intron XP_016868115.1
XM_017012627.1 Intron XP_016868116.1
XM_017012628.1 Intron XP_016868117.1
XM_017012629.1 Intron XP_016868118.1
XM_017012630.1 Intron XP_016868119.1
XM_017012631.1 Intron XP_016868120.1
XM_017012632.1 Intron XP_016868121.1
XM_017012633.1 Intron XP_016868122.1
XM_017012634.1 Intron XP_016868123.1
XM_017012635.1 Intron XP_016868124.1
XM_017012636.1 Intron XP_016868125.1
XM_017012637.1 Intron XP_016868126.1
XM_017012638.1 Intron XP_016868127.1
XM_017012639.1 Intron XP_016868128.1
XM_017012640.1 Intron XP_016868129.1
XM_017012641.1 Intron XP_016868130.1
XM_017012642.1 Intron XP_016868131.1
Gene
STEAP2
Gene Name
STEAP2 metalloreductase
There are no transcripts associated with this gene.

View Full Product Details