Product Details

SNP ID
rs16827358
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:40533803 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATAACAAATATTGTCAAGGTTCAT[A/G]AAGCAGTATGTTAGGTATTCTTTTT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF684 PubMed Links

Gene Details

Gene
ZNF684
Gene Name
zinc finger protein 684
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152373.3 Intron NP_689586.3
XM_011540671.2 Intron XP_011538973.1
XM_011540672.2 Intron XP_011538974.1
XM_011540673.2 Intron XP_011538975.1
XM_017000290.1 Intron XP_016855779.1
XM_017000291.1 Intron XP_016855780.1
XM_017000292.1 Intron XP_016855781.1

View Full Product Details