Product Details

SNP ID
rs17592236
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.13:40557795 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAAAATCCCTCGTTTGACAAAGGA[C/T]CATTGCTTTAGATGCAGATCTCCCT
Phenotype
MIM: 136533
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FOXO1 PubMed Links

Gene Details

Gene
FOXO1
Gene Name
forkhead box O1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002015.3 3607 UTR 3 NP_002006.2
XM_011535008.2 3607 UTR 3 XP_011533310.1
XM_011535010.2 3607 UTR 3 XP_011533312.1

View Full Product Details