Product Details

SNP ID
rs17177486
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:203341719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTTAGGGAGGTATAGAAGTGACT[C/T]CTGGTGTCCTTTCACCCTCAGACAC
Phenotype
MIM: 600245
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FMOD PubMed Links

Gene Details

Gene
FMOD
Gene Name
fibromodulin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002023.4 2159 UTR 3 NP_002014.2

View Full Product Details