Product Details

SNP ID
rs16860432
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47598898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTATTCATTGAATGTCTAAGTATGT[C/G]CCAGATATCATATACAGATATGTGA
Phenotype
MIM: 605236
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ATP10D PubMed Links

Gene Details

Gene
ATP10D
Gene Name
ATPase phospholipid transporting 10D (putative)
There are no transcripts associated with this gene.

Gene
CORIN
Gene Name
corin, serine peptidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278585.1 Intron NP_001265514.1
NM_001278586.1 Intron NP_001265515.1
NM_006587.3 Intron NP_006578.2

View Full Product Details