Product Details
- SNP ID
-
rs17075291
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:184090745 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGTACAGGATTGTGGCCACAGACAC[C/T]GAGGATGGCTCCTGGGTGTGGACAG
- Phenotype
-
MIM: 616983
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ENPP6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114488141] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ENPP6
- Gene Name
- ectonucleotide pyrophosphatase/phosphodiesterase 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_153343.3 |
1897 |
UTR 3 |
|
|
NP_699174.1 |
View Full Product Details