Product Details

SNP ID
rs34043510
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38286896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAAACAGCGCCTCCTCAGGTCTCC[C/T]TGAGCCAATCACTTGAAAAGGGCAA
Phenotype
MIM: 607083
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WHSC1L1 PubMed Links

Gene Details

Gene
WHSC1L1
Gene Name
Wolf-Hirschhorn syndrome candidate 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017778.2 Intron NP_060248.2
NM_023034.1 Intron NP_075447.1

View Full Product Details