Product Details

SNP ID
rs16956811
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:57410090 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCAGCCACAGCTGTCAAATGCAAC[G/T]GAAGATGATGAAGAGCTGCCGCCCC
Phenotype
MIM: 601520
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CCL17 PubMed Links

Gene Details

Gene
CCL17
Gene Name
C-C motif chemokine ligand 17
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002987.2 Intron NP_002978.1
XM_011523256.2 Intron XP_011521558.1
XM_017023530.1 Intron XP_016879019.1

View Full Product Details