Product Details

SNP ID
rs17854186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:74142324 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTTCTCTTCTCCTTAAATTCTTCT[G/T]ATTCAATTTTAATTTCATACATCGC
Phenotype
MIM: 613183
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BOLA3 PubMed Links

Gene Details

Gene
BOLA3
Gene Name
bolA family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001035505.1 Intron NP_001030582.1
NM_212552.2 Intron NP_997717.2
Gene
BOLA3-AS1
Gene Name
BOLA3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details