Product Details

SNP ID
rs72552795
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.7:99760842 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CCTGTGTTTCTTTACAAGGTTTGAA[-/G]GGAGAAGTTCTGAAGGACTCTGATT
Phenotype
MIM: 124010
Polymorphism
-/G, Insertion/deletion
Allele Nomenclature
Literature Links
CYP3A4 PubMed Links

Gene Details

Gene
CYP3A4
Gene Name
cytochrome P450 family 3 subfamily A member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202855.2 1496 Frame Shift InDel NP_001189784.1
NM_017460.5 1496 Frame Shift InDel NP_059488.2
XM_017011780.1 1496 Intron XP_016867269.1

View Full Product Details