Product Details

SNP ID
rs16962873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:84147418 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCGGTTGAATTGGCCAATTTTGAA[C/T]GACCTTTAAAAAAATTATTCATTTG
Phenotype
MIM: 613190
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAAF1 PubMed Links

Gene Details

Gene
DNAAF1
Gene Name
dynein axonemal assembly factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318756.1 Intron NP_001305685.1
NM_178452.5 Intron NP_848547.4
XM_006721129.2 Intron XP_006721192.1
XM_011522853.2 Intron XP_011521155.1
XM_011522854.2 Intron XP_011521156.1
XM_011522855.2 Intron XP_011521157.1
XM_011522857.2 Intron XP_011521159.1
XM_011522858.2 Intron XP_011521160.1
XM_017022918.1 Intron XP_016878407.1
XM_017022919.1 Intron XP_016878408.1
XM_017022920.1 Intron XP_016878409.1
XM_017022921.1 Intron XP_016878410.1
XM_017022922.1 Intron XP_016878411.1
Gene
HSDL1
Gene Name
hydroxysteroid dehydrogenase like 1
There are no transcripts associated with this gene.

View Full Product Details