Product Details

SNP ID
rs16976743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43732259 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTTAAGCTATTTATCTTTCCAA[C/T]TTTTCTCTTTGAAGTAAGATGAGGA
Phenotype
MIM: 613176
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SMG9 PubMed Links

Gene Details

Gene
SMG9
Gene Name
SMG9, nonsense mediated mRNA decay factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005259057.2 Intron XP_005259114.1
XM_011527113.1 Intron XP_011525415.1
XM_011527114.1 Intron XP_011525416.1
XM_011527115.1 Intron XP_011525417.1
XM_011527116.1 Intron XP_011525418.1
XM_017026988.1 Intron XP_016882477.1

View Full Product Details