Product Details
- SNP ID
-
rs17037420
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
31
- Location
-
Chr.1:11807759 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATAAAGAAGGTAGAGAAAAGCCT[A/G]TAACTCCATTCATTAAAAGATGACT
- Phenotype
-
MIM: 602726
MIM: 607093
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CLCN6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76351797] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CLCN6
- Gene Name
- chloride voltage-gated channel 6
- Gene
- MTHFR
- Gene Name
- methylenetetrahydrofolate reductase (NAD(P)H)
There are no transcripts associated with this gene.
View Full Product Details