Product Details

SNP ID
rs16831216
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:134965311 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCACACTTACATTTACTTTTAAAA[C/T]TCAGAAGCATTAAAATCCAGAATTG
Phenotype
MIM: 603862
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNT2 PubMed Links

Gene Details

Gene
CCNT2
Gene Name
cyclin T2
There are no transcripts associated with this gene.

Gene
MAP3K19
Gene Name
mitogen-activated protein kinase kinase kinase 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018044.2 Intron NP_001018054.1
NM_001018046.2 Intron NP_001018056.1
NM_001018047.2 Intron NP_001018057.1
NM_001282883.1 Intron NP_001269812.1
NM_001321177.1 Intron NP_001308106.1
NM_025052.4 Intron NP_079328.3
XM_011511891.2 Intron XP_011510193.1
XM_011511892.1 Intron XP_011510194.1
XM_011511894.2 Intron XP_011510196.1
XM_011511896.2 Intron XP_011510198.1
XM_011511897.2 Intron XP_011510199.1
XM_017005003.1 Intron XP_016860492.1
XM_017005004.1 Intron XP_016860493.1
XM_017005005.1 Intron XP_016860494.1
XM_017005006.1 Intron XP_016860495.1
XM_017005007.1 Intron XP_016860496.1

View Full Product Details