Product Details

SNP ID
rs17331590
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:69219666 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTGTACTCACCTATTTAGAATTCA[A/G]ATCAGGTTCGTGATTGCGCTCTTAA
Phenotype
MIM: 605607 MIM: 611996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CENPH PubMed Links

Gene Details

Gene
CENPH
Gene Name
centromere protein H
There are no transcripts associated with this gene.

Gene
MRPS36
Gene Name
mitochondrial ribosomal protein S36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033281.5 Intron NP_150597.1

View Full Product Details